Filter papers for soil analysis
Leverandør: Ahlstrom
Om denne vare
These papers are used to determine the presence of trace elements and nutrients in soil samples to optimise plant growth.
4 tilgængelige muligheder nedenfor
Product Details & Documents
These papers are used to determine the presence of trace elements and nutrients in soil samples to optimise plant growth.
Specifications for Product Family
Specifikationer
- Catalog No:
- 516-0776
- BINZ146029
- BINZ146030
- BINZ146032
- Kvalitet:
- 132
- 132
- 132
- 132
- Vægt:
- 80 g/m²
- 80 g/m²
- 80 g/m²
- 80 g/m²
- Filtreringshastighed:
- 450 s (Herzberg)
- 450 s (Herzberg)
- 450 s (Herzberg)
- 450 s (Herzberg)
Specifikationer
Product Family Options
Circles
Produktinformation
- KvalitetStørrelseTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.516-0776Kvalitet132Vægt80 g/m²Filtreringshastighed450 s (Herzberg)
Folded filters
Produktinformation
- KvalitetStørrelseTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.BINZ146029Kvalitet132Vægt80 g/m²Filtreringshastighed450 s (Herzberg) - Specifikationer:
More
Specifications
Cat. No.BINZ146030Kvalitet132Vægt80 g/m²Filtreringshastighed450 s (Herzberg) - Specifikationer:
More
Specifications
Cat. No.BINZ146032Kvalitet132Vægt80 g/m²Filtreringshastighed450 s (Herzberg)