8-Bromisoquinolin
Glass bottle for solids, 1 g
Katalog # ACTIAS1288G1
Marketsource
Leverandør: ACTIVATE SCIENTIFIC
8-Bromisoquinolin
Katalog # ACTIAS1288G1
Leverandør: ACTIVATE SCIENTIFIC
Marketsource
CAS nummer: 63927-22-0
Gå til:
Product Details & Documents
- CAS nummer:63927-22-0
- Formel:C₉H₆BrN
- MDL nummer:MFCD04973298
- Molecular Weight:208.0600 g/mol
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



