Dextran, M.W. ~ 500,000
Glass bottle for solids, 100 g
Katalog # SERA18696.01
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
- CAS nummer:9004-54-0
- Formel:(C₆H₁₀O₅)n
- MDL nummer:MFCD00130935
- EINECS:232-677-5
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 g



