Orders LinkContactUsLinkComponent
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), 1
Katalog # PIER28355
Leverandør:   Thermo Fisher Scientific
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
Katalog # PIER28355
CAS nummer:  
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), 1
CAS nummer:  
Price
1.460,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
Specifikationer
  • Pakn:
    1
  • Beskrivelse:
    TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)
Specifikationer