Touch screen gloves, GL16
Leverandør: Portwest
Om denne vare
These knitted gloves are made of 100% acrylic fibres. The black acrylic parts are hardwearing while the grey acrylic with metallic component allows for precise movements on a touch screen device.
- High dexterity
- Grey acrylic fabric on thumb and two fingers
2 tilgængelige muligheder nedenfor
Product Details & Documents
These knitted gloves are made of 100% acrylic fibres. The black acrylic parts are hardwearing while the grey acrylic with metallic component allows for precise movements on a touch screen device.
- High dexterity
- Grey acrylic fabric on thumb and two fingers
Specifications for Product Family
Specifikationer
- Catalog No:
- 112-2951
- 112-2950
- Model:
- GL16
- GL16
- Ambidextrous/Hånd-specifik:
- Hånd-specifik
- Hånd-specifik
- Engangs/Genbrugelig:
- Genanvendelig
- Genanvendelig
- Manchet:
- Strikket
- Strikket
Specifikationer
Product Family Options
Produktinformation
- Handske størrelseFarveTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.112-2951ModelGL16Ambidextrous/Hånd-specifikHånd-specifikEngangs/GenbrugeligGenanvendeligManchetStrikket - Specifikationer:
More
Specifications
Cat. No.112-2950ModelGL16Ambidextrous/Hånd-specifikHånd-specifikEngangs/GenbrugeligGenanvendeligManchetStrikket