Wet mop handles, Invader
129-0752
- Colour:blå
- Beskrivelse:Mop handle, Invader
- Length:1520 mm
- Environmentally Preferable:
- Pakn:1
Wet mop handle made of fibreglass with a plastic head. Connection type: Side gate. For use with 1 and 5" headband mops or use with 1" headbands only.