Fleece jacket, Merida 612Z S.E.P.P.
: SIOEN
Protective, single colour jacket made of 100% soft double-sided polyester fleece providing good cold protection. With anti-pilling treatment and stitched seams.
- Fixed windbreaker lining in front and back
- Straight collar, zip closure with chin protection and extra long zip puller
- One inset chest pocket and two inset front pockets with zip closure
- Round inset sleeves, elasticated cuffs
- Drawstring in hem
Can be zipped into the rain jacket 608Z.
Weight: 285 g/m²
Certifications: EN 14058: 2004 (1XXXX)
Protective, single colour jacket made of 100% soft double-sided polyester fleece providing good cold protection. With anti-pilling treatment and stitched seams.
- Fixed windbreaker lining in front and back
- Straight collar, zip closure with chin protection and extra long zip puller
- One inset chest pocket and two inset front pockets with zip closure
- Round inset sleeves, elasticated cuffs
- Drawstring in hem
Can be zipped into the rain jacket 608Z.
Weight: 285 g/m²
Certifications: EN 14058: 2004 (1XXXX)
- Catalog No:
- 113-07428
- 113-07429
- 113-07430
- 113-07431
- 113-07432
- 113-07433
- 113-07434
- 113-07435
- 113-07436
- 113-07437
- 113-07438
- 113-07439
- 113-07440
- 113-07441
- 113-07449
- 113-07450
- 113-07451
- 113-07452
- 113-07453
- 113-07454
- 113-07455
- 113-07442
- 113-07443
- 113-07444
- 113-07445
- 113-07446
- 113-07447
- 113-07448
- Model:
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- 612Z Merida
- Outfit applikationer:
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- Risk area:
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Køn:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Lukkestørrelse:
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Stof:
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- 100% soft double-sided polyester
- Vægt:
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
Product Family Options
- FarveTøj størrelse
Specifications
Cat. No.113-07428Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07429Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07430Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07431Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07432Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07433Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07434Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07435Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07436Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07437Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07438Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07439Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07440Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07441Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07449Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07450Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07451Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07452Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07453Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07454Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07455Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07442Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07443Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07444Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07445Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07446Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07447Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²Specifications
Cat. No.113-07448Model612Z MeridaOutfit applikationerGeneral PurposeRisk areaColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% soft double-sided polyesterVægt285 g/m²