Disposable particle filtering respirators, FFP1/FFP2/FFP3, Smart Pocket
Catalog # 111-1144
Supplier: Moldex
Specifications
- Model:2575
- Seq:03
- Størrelse:U*
- Pakn:10
- Med ventil:+
Specifications
About this item
Folded particulate filtering masks with or without valves.
- Easily unfolds in a quick one step process
- Comfortable nose seal
- DuraMesh® - strong and durable structure, keeps the mask in shape
- ActivForm® - no nose clip needed
- Loop strap with clip for quick, simple and safe on and off
- Individually packaged
EN 149:2001+A1:2009