Freezer thermometer
Calibrated thermometer with holder, −35...+25 °C, 265 mm
Katalog # HECH43599010
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
This freezer thermometer made of plastic is supplied with holder and factory certificate. Officially verified.
Specifikationer
- Beskrivelse:Calibrated thermometer with holder
- Længde:265 mm
- Område:−35...+25 °C