Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Leverandør: AGILENT HEWLETT PACKARD
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Leverandør: AGILENT HEWLETT PACKARD
Certifikater
Gå til:
Om denne vare
33 tilgængelige muligheder nedenfor
Product Details & Documents
Product Family Options
Flow cell
Produktinformation
- TilgængelighedPrice
- HEWLG1315-60011Semi-micro flow cell, 6 mm, 5 μl, 120 bar for G1315A/B, G1365A/BSemi-micro flow cell, 6 mm, 5 μl, 120 bar for G1315A/B, G1365A/BLog ind for tilgængelighed1 * 1 Items10.970,00 kr
- HEWLG1315-60012Standard flow cell, 10 mm, 13 μl, 120 bar for G1315A/B, G1365A/BStandard flow cell, 10 mm, 13 μl, 120 bar for G1315A/B, G1365A/BLog ind for tilgængelighed1 * 1 Items10.970,00 kr
- HEWLG1315-60015Micro high-pressure flow cell, 6 mm, 1,7 μl, 400 bar for G1315A/B, G1365A/BMicro high-pressure flow cell, 6 mm, 1,7 μl, 400 bar for G1315A/B, G1365A/BLog ind for tilgængelighed1 * 1 Items29.250,00 kr
- HEWLG1315-60016Preparative flow cell, 3 mm, 120 bar, stainless steel for G1315A/B, G1365A/BPreparative flow cell, 3 mm, 120 bar, stainless steel for G1315A/B, G1365A/BLog ind for tilgængelighed1 * 1 Items12.610,00 kr
- HEWLG1315-60017Preparative flow cell, 0,3 mm, 20 bar, quartz
- HEWLG1315-60018Preparative flow cell, 0,06 mm, 20 bar, quartz
- HEWLG1315-60022Standard flow cell with RFID tag, 10 mm, 13 μl, 120 bar for G1315C/D, G1365C/DStandard flow cell with RFID tag, 10 mm, 13 μl, 120 bar for G1315C/D, G1365C/DLog ind for tilgængelighed1 * 1 Items16.570,00 kr
- HEWLG1315-60024Micro flow cell, RFID, 3 mm, 2 μl, 120 bar for G1315C/D, G1365C/DMicro flow cell, RFID, 3 mm, 2 μl, 120 bar for G1315C/D, G1365C/DLog ind for tilgængelighed1 * 1 Items20.150,00 kr
- HEWLG1315-60025Semi-micro flow cell, RFID, 6 mm, 5 μl, 120 bar for G1315C/D, G1365C/DSemi-micro flow cell, RFID, 6 mm, 5 μl, 120 bar for G1315C/D, G1365C/DLog ind for tilgængelighed1 * 1 Items18.280,00 kr
- HEWLG1315-6871680 nl flow cell, 6 mm, 50 bar
- HEWLG1315-68724500 nl flow cell, 10 mm, 50 bar
Repair kits
Produktinformation
- TilgængelighedPrice
- HEWLG1315-68712Repair kit for standard flow cell, standard flow cell with RFID tag and preparative flow cellRepair kit for standard flow cell, standard flow cell with RFID tag and preparative flow cellLog ind for tilgængelighed1 * 1 Items3.940,00 kr
Capillaries for DAD/MWD flow cell
Produktinformation
- TilgængelighedPrice
- HEWLG1315-87302Outlet capillary, 0,17 mm ID, 200 mm long for standard flow cell, standard flow cell with RFID tagOutlet capillary, 0,17 mm ID, 200 mm long for standard flow cell, standard flow cell with RFID tagLog ind for tilgængelighed1 * 1 Items1.060,00 kr
- HEWLG1315-87321Inlet capillary with heat exchanger, 0,17 mm ID, 590 mm long for standard flow cell, standard flow cell with RFID tagInlet capillary with heat exchanger, 0,17 mm ID, 590 mm long for standard flow cell, standard flow cell with RFID tagLog ind for tilgængelighed1 * 1 Items1.060,00 kr
- HEWLG1315-87325Inlet capillary with heat exchanger, 0,12 mm ID, 290 mm long for micro high-pressure flow cellInlet capillary with heat exchanger, 0,12 mm ID, 290 mm long for micro high-pressure flow cellLog ind for tilgængelighed1 * 1 Items1.060,00 kr
80 nl and 500 nl flow cell supplies
Produktinformation
- TilgængelighedPrice
- HEWL5022-2146Union adjustment tool
- HEWL5063-6592Ferrule, 1/32" and stainless steel lock ring, lite touchFerrule, 1/32" and stainless steel lock ring, lite touchLog ind for tilgængelighed1 * 10 Items679,00 kr
500 nl flow cell and replacement parts
Produktinformation
- TilgængelighedPrice
- HEWLG1315-68715Sealing kit (includes torque adapter, 2 cell seal assemblies, 5 lite touch front and back ferrules)Sealing kit (includes torque adapter, 2 cell seal assemblies, 5 lite touch front and back ferrules)Log ind for tilgængelighed1 * 1 Items
- HEWLG1315-80001Quartz cell body, 10 mm
- HEWLG1315-87323Fused silica/PEEK capillary, 50 μm ID, 40 cm long, inletFused silica/PEEK capillary, 50 μm ID, 40 cm long, inletLog ind for tilgængelighed1 * 1 Items1.390,00 kr
- HEWLG1315-87328Fused silica/PEEK capillary, 50 μm ID, 12 cm long, outletFused silica/PEEK capillary, 50 μm ID, 12 cm long, outletLog ind for tilgængelighed1 * 1 Items1.270,00 kr
- HEWLG1315-87333Fused silica/PEEK capillary, 100 μm ID, 30 cm long, inletFused silica/PEEK capillary, 100 μm ID, 30 cm long, inletLog ind for tilgængelighed1 * 1 Items1.230,00 kr
- HEWLG1315-87338Fused silica/PEEK capillary, 100 μm ID, 12 cm long, outletFused silica/PEEK capillary, 100 μm ID, 12 cm long, outletLog ind for tilgængelighed1 * 1 Items1.230,00 kr
80 nl flow cell and replacement parts
Produktinformation
- TilgængelighedPrice
- HEWLG1315-68725Sealing kit for 80 nl flow cell (includes torque adapter, 2 cell seal assemblies, 5 lite touch front and back ferrules and 5 sleeves for 360 μm OD capillaries)Sealing kit for 80 nl flow cell (includes torque adapter, 2 cell seal assemblies, 5 lite touch front and back ferrules and 5 sleeves for 360 μm OD capillaries)Log ind for tilgængelighed1 * 1 Items
- HEWLG1315-80002Quartz cell body, 80 nl, 6 mm path length
- HEWLG1315-87313Fused silica/PEEK capillary, 25 μm ID, 20 cm long, inletFused silica/PEEK capillary, 25 μm ID, 20 cm long, inletLog ind for tilgængelighed1 * 1 Items1.230,00 kr
- HEWLG1315-87318Fused silica/PEEK capillary, 25 μm ID, 60 cm long, outletFused silica/PEEK capillary, 25 μm ID, 60 cm long, outletLog ind for tilgængelighed1 * 1 Items1.390,00 kr
Preparative flow cells and replacement parts
Produktinformation
- TilgængelighedPrice
- HEWL5042-6443Back pressure regulator, 100 psi for G1315B/DAD/MWDBack pressure regulator, 100 psi for G1315B/DAD/MWDLog ind for tilgængelighed1 * 1 Items1.780,00 kr
- HEWLG1315-27705Cell housing
- HEWLG1315-80004Quartz body, 0,3 mm
- HEWLG1315-87305Connecting capillary, stainless steel, 0,5 mm, 250 mmConnecting capillary, stainless steel, 0,5 mm, 250 mmLog ind for tilgængelighed1 * 1 Items843,00 kr
Seals
Produktinformation
- TilgængelighedPrice
- HEWLG1315-68710Front seals, 1,3 mm hole, PTFE
- HEWLG1315-68711Back seals, 2,3 mm hole, PTFE
Repair kits
Produktinformation
- Katalog #BeskrivelseTilgængelighedPrice
- Repair Kit for semi-micro flow cell and RFID (6 mm, 5μl, 120bar), micro flow cell, RFID, (3 mm, 2μl, 120bar)Log ind for tilgængelighed1 * 1 Items3.940,00 kr
Capillaries for DAD/MWD flow cell
Produktinformation
- Katalog #BeskrivelseTilgængelighedPrice
- DAD heat exchanger capillary, 1 end pre-swaged, male to male, 310 mm, I.Ø 0,12 mmLog ind for tilgængelighed1 * 1 Items1.000,00 kr
80 nl and 500 nl flow cell supplies
Produktinformation
- Katalog #BeskrivelseTilgængelighedPrice
- 1/16" stainless steel fittings, male, 4 mm, fitting type BLog ind for tilgængelighed1 * 10 Items687,00 kr
Preparative flow cells and replacement parts
Produktinformation
- Katalog #BeskrivelseTilgængelighedPrice