Repetitive pipette, Stepper™ 416
Katalog # SOCO416.5000
Leverandør: Socorex Isba
Specifikationer
- Kapacitet:10 - 5000 µl
- Beskrivelse:Adjustable repetitive pipette, Stepper™ 416
- Seq:01
- Environmentally Preferable:
- Pakn:1
Specifikationer
Om denne vare
Adjustable pipette intended for reliable dispensing and high performance results. Selected materials offer outstanding shock resistance and compact shape greatly limit internal contamination. The pipette provides up to 73 doses within a large selection of 53 different volumes.
- Volumes range from 10 to 5000 µl
- Unique trigger action (Activation with 4-finger instead of thumb) eliminates thumb fatigue
- Setting knobs bear clear indication of volumes and number of aliquots
- Only three positive displacement syringes, colour coded
- Self-locking mechanism
Syringes are available non sterile (supplied in bulk packages) or sterile (bioproof™ version, individually wrapped). Selection packs include 20 syringes of each volume.
Ordering information: Supplied with colour coded selector buttons, adapter, Ecostep™ syringes.