Lactodensimeters
Leverandør: GERBER FUNKE
Gå til:
Om denne vare
This lactodensimeter according to Gerber is designed with a thermometer.
- Thermometer included in stem or body
- Standard model with negative scale
- Calibrated temperature of 20 °C
2 tilgængelige muligheder nedenfor
Product Details & Documents
This lactodensimeter according to Gerber is designed with a thermometer.
- Thermometer included in stem or body
- Standard model with negative scale
- Calibrated temperature of 20 °C
Product Family Options
Produktinformation
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Replacement hanging cylinder (210 mm)