SPE columns, CHROMABOND® CN
Leverandør: MACHEREY-NAGEL
Om denne vare
CHROMABOND® CN is a cyanopropyl modified silica phase for solid phase extraction.
- Polar to mid-polar phase
- Weak hydrophobic and selective interactions by the high electron density of the CN group
- Suitable for polar and aromatic compounds
Recommended for substances with a high electron density like cyclosporins or for polar carbohydrates.
2 tilgængelige muligheder nedenfor
Product Details & Documents
CHROMABOND® CN is a cyanopropyl modified silica phase for solid phase extraction.
- Polar to mid-polar phase
- Weak hydrophobic and selective interactions by the high electron density of the CN group
- Suitable for polar and aromatic compounds
Recommended for substances with a high electron density like cyclosporins or for polar carbohydrates.
Specifications for Product Family
Specifikationer
- Catalog No:
- MANA730061
- MANA730063
- Kulstofbelastning:
- 5,5%
- 5,5%
- Endcapped:
- Ingen
- Ingen
- pH-område:
- 2 - 8
- 2 - 8
- Surface area:
- 500 m²/g
- 500 m²/g
Specifikationer
Product Family Options
Produktinformation
- FaseKolonnematerialePorestørrelsePartikelstørrelseKapacitetLejevægtTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.MANA730061Kulstofbelastning5,5%EndcappedIngenpH-område2 - 8Surface area500 m²/g - Specifikationer:
More
Specifications
Cat. No.MANA730063Kulstofbelastning5,5%EndcappedIngenpH-område2 - 8Surface area500 m²/g