Orders LinkContactUsLinkComponent
Triangular bandage, Mitella
Triangular bandage, Mitella
Triangular bandage, 960×960×136 mm
Catalog # 118-0249
Triangular bandage, Mitella
Catalog # 118-0249
Triangular bandage, Mitella
Triangular bandage, Mitella
Triangular bandage, 960×960×136 mm
Price
56,40 kr
Log in for availability
1 * 1 Items
Product Details & Documents
This bandage is tied around the neck to support an injured arm. It can also be wound around the head to keep a head wound dressing in place or rolled up for use as a tourniquet.
  • Consists of two triangular bandages and six safety pins
  • Made of white spunbonded polypropylene
  • Packed in a printed box
Specifications
  • Beskrivelse:
    Triangular bandage
  • B×D×H:
    960×960×136 mm