Orders LinkContactUsLinkComponent
Hoffmann clips, heavy duty
Hoffmann clips, heavy duty
40 mm
Catalog # MURRL142/02
Supplier:   MURRAY S
Hoffmann clips, heavy duty
Catalog # MURRL142/02
Supplier:  MURRAY S
Hoffmann clips, heavy duty
40 mm
Price
392,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Heavy duty clip fitted with nickel plated brass tommy screw and hinged bottom bar.
  • Spanning width = Width of gate
Specifications
  • Spændvidde:
    40 mm