Cuvettes for Fibrintimer®
1,3 ml, PS, 11,9×34,5 mm
Katalog # KART2630
Leverandør: KARTELL
Gå til:
Product Details & Documents
Polystyrene cuvettes for Fibrintimer used to determine the coagulation-time of blood.
Certifications: IVD (in vitro diagnostic) certified.
Delivery information: Supplied with 500 metallic mixers.
Specifikationer
- Diameter×H:11,9×34,5 mm
- Volumen:1,3 ml
- Materiale:PS