Cuvettes for Olli-C-Analyzer®
4 ml, PS, 11,85×50,80 mm
Katalog # KART2618
Leverandør: KARTELL
Gå til:
Product Details & Documents
These cylindrical cells are specifically used in conjunction with Olli-C-Analyzer.
- Anti-shock polystyrene-moulding design
- Highly resistant and dust-proof technology
- Quality optical material suitable for IVD applications
Packaging: In polystyrene dust proof boxes of 192 pieces.
Specifikationer
- Diameter×H:11,85×50,80 mm
- Volumen:4 ml
- Materiale:PS