Bandage scissors, bent
Universal bandage scissors, saw edge, 175 mm
Katalog # MURR1917/05
Leverandør: MURRAY S
Gå til:
Product Details & Documents
Universal bandage scissors made of stainless steel. With black autoclavable handle.
Specifikationer
- Model:Universal bandage scissors, saw edge
- Længde:175 mm