Orders LinkContactUsLinkComponent
Sockets for rods without thread
Sockets for rods without thread
Supplier:   BOCHEM
Sockets for rods without thread
Supplier:  BOCHEM
Certificates
Sockets for rods without thread
Sockets for rods without thread
2 Options Available Below
Product Details & Documents
These sockets are available in two materials: 18/10 stainless steel or green coated steel.
Suitable for rods with a diameter of 12 - 13 mm.
Product Family Options
Product Information
  • Ø
    Beskrivelse
    Materiale
    Availability
    Price
  • 13,2 mm
    Socket without thread, for 12 - 13 mm Ø rods
    Rustfrit stål 18/10
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 211,00 kr
    211,00 kr
    1 * 1 Items
       211,00 kr
  • 13,2 mm
    Socket without thread, green coated, for 12 - 13 mm Ø rods
    Stål
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 125,00 kr
    125,00 kr
    1 * 1 Items
       125,00 kr