Cell scrapers
Celleskraber med blad med to positioner, 250 mm
Catalog # SARS83.3951
Marketsource
Supplier: SARSTEDT
Jump to:
Product Details & Documents
Cell scraper with soft, flexible blade for gentle removal of monolayer cultures, minimising cell damage.
- Easy and complete separation of monolayer due to extra thin and flexible blade
- When downward pressure is applied to the handle the blade can remove all cells in its path in one pass
- Sterilised by gamma irradiation, individually wrapped
Specifications
- Beskrivelse:Celleskraber med blad med to positioner
- Seq:01
- Samlet længde:250 mm