Aerosol analysis monitors
Om denne vare
The PS monitor is used for sample collection and particle analysis. The PP monitor is used for microscopic particle analysis and sample collection for HPLC and UV detection.
Membrane: mixed cellulose esters with cellulose support pad; housing: PS or carbon filled PP.
3 tilgængelige muligheder nedenfor
Product Details & Documents
The PS monitor is used for sample collection and particle analysis. The PP monitor is used for microscopic particle analysis and sample collection for HPLC and UV detection.
Membrane: mixed cellulose esters with cellulose support pad; housing: PS or carbon filled PP.
Specifications for Product Family
Specifikationer
- Catalog No:
- MILFMAWP025A0
- MILFMAWP025AC
- MILFAP1002500
- Porestørrelse:
- 0,8/5,0 µm
- 0,8/5,0 µm
- Materiale:
- PS
- PP
- Filtreringsareal:
- 4 cm²
- 4 cm²
- Ø:
- 25 mm
- 25 mm
- Indløb/udløb forbindelser:
- Female Luer slip/female Luer slip
- Hose barb/-
Specifikationer
Product Family Options
Produktinformation
- TilgængelighedPrice
- MILFMAWP025A03-piece PS monitor with 16 mm center ring and female Luer slip vacuum connection3-piece PS monitor with 16 mm center ring and female Luer slip vacuum connectionLog ind for tilgængelighed1 * 50 Items2.210,00 krSpecifikationer:
More
Specifications
Cat. No.MILFMAWP025A0Porestørrelse0,8/5,0 µmMaterialePSFiltreringsareal4 cm²Ø25 mmIndløb/udløb forbindelserFemale Luer slip/female Luer slip - MILFMAWP025AC3-piece carbon filled PP conductive monitor with 50 mm extension cowl3-piece carbon filled PP conductive monitor with 50 mm extension cowlLog ind for tilgængelighed1 * 50 Items2.080,00 krSpecifikationer:
More
Specifications
Cat. No.MILFMAWP025ACPorestørrelse0,8/5,0 µmMaterialePPFiltreringsareal4 cm²Ø25 mmIndløb/udløb forbindelserHose barb/- - MILFAP1002500Absorbent pad, 25 mmSpecifikationer:
More
Specifications
Cat. No.MILFAP1002500PorestørrelseMaterialeFiltreringsarealØIndløb/udløb forbindelser