Orders LinkContactUsLinkComponent
DIALYZER-Enhance™
DIALYZER-Enhance™
DIALYZER-Enhance™, for improved dialysis time and rates, 50 mL
Catalog # 786-627
Supplier:   G-Biosciences
DIALYZER-Enhance™
Catalog # 786-627
Supplier:  G-Biosciences
CAS Number:  
DIALYZER-Enhance™
DIALYZER-Enhance™
DIALYZER-Enhance™, for improved dialysis time and rates, 50 mL
CAS Number:  
Price
927,00 kr
Log in for availability
1 * 50 mL
Product Details & Documents
For improved dialysis time and rates.
A product that when added to the dialysis buffer shifts the equilibrium resulting in the increased removal of a wide range of small molecules. The DIALYZER-Enhance™ consists of unreactive reagents that will not interfere or modify your reagents and will not cross the dialysis membrane, ensuring a pure, clean sample at the end of dialysis.
Documents
Specifications
Specifications
  • Pakn:
    50 mL
  • Beskrivelse:
    DIALYZER-Enhance™, for improved dialysis time and rates