Orders LinkContactUsLinkComponent
Half Mask HF-800 SeriesFeaturecard.png
3M Secure Click Reusable Half Mask HF-800 Series
Designed with smart and intuitive features 3M Secure Click Half Mask Reusable Respirator HF-800 Series is simple, comfortable and reliable. Available in three sizes: small, medium, and large, our respirators all come with an optional speaking diaphragm.
Reusable Half Mask HF-800 Series | Avantor
Half Mask HF-800 Column.png
3M Secure Click Reusable Half Mask HF-800 Series
Designed with smart and intuitive features 3M Secure Click Half Mask Reusable Respirator HF-800 Series is simple, comfortable and reliable. Available in three sizes: small, medium, and large, our respirators all come with an optional speaking diaphragm. The new 3M Secure Click Series features a quick, simple cartridge/filter connection. It works just like a seat belt. Simply align the connections and push until you hear a click and feel confident that filters and cartridges are installed properly.