Orders LinkContactUsLinkComponent
European standards for occupational and safety shoes | Avantor
European standards for occupational and safety shoes and footwear
Standards
EN ISO 20345: Safety footwear (200 J toecap) with improved slipping resistance - Category S
EN ISO 20345:2011*: New updated standard for safety footwear - Category S
EN ISO 20346: Protective footwear (100 J toecap) - Category P
EN ISO 20347: Occupational footwear (without toecap) - Category O
The conformity to the norm is certified by a CE certificate.
European standards
EN ISO 20345 Safety footwear (200 J toecap) SB S1 S1P S2 S3
EN ISO 20346 Protective footwear (100 J toecap) PB P1 - P2 P3
EN ISO 20347 Occupational footwear (without toecap OB O1 - O2 O3
Covered risks
Fundamentals risks** X X X X X
Enclosed back - X X X X
Profiled outer sole O X X X X
Professional use category
A Anti-static O X X X X
E Shock absorbing heel O X X X X
WRU Water resistant upper O - - X X
P Puncture resistant sole O - X - X
C Electrical resistance, conductivity O O O O O
HI Heat insulation O O O O O
CI Cold insulation O O O O O
HRO Contact-heat resistant outer sole O O O O O
AN Ankle bone protection O O O O O
WR Water resistance of the entire shoe O O O O O
M Metatarsal protection O O O O O
X = obligatory
O = optional
* All products must be certified and manufactured in accordance with the new standard. As from 30th June 2013, manufacturers are obliged to certify, produce and commercialise all products in accordance with EN ISO 20345:2011. Distributors should purchase only products certified to the 2011 version of the standard; however they are permitted to continue the sale of products certified to the previous standards already in their stocks, with no limit on the end date.
** The fundamental risks are defined as follows: The quality and the performance of the incorporated materials (leathers, lining, soles, etc.), tearing flexing and abrasing resistance, permeability to water and vapour, upper sole grip and non-slip properties of the sole.
Outsole slip resistance
SRA
Test ground: Ceramic
Lubricant: Sodium lauryl sulfate
>0,32 flat footwear
>0,28 footwear with a 7° heel
SRB
Test ground: Steel
Lubricant: Glycerine
>0,18 flat footwear
>0,13 footwear with a 7° heel
SRC
SRC = SRA+SRB
ESD (Electro Static Discharge)
ESD shoes protect electrostatically endangered components in sensitive workspaces and prevent sparks and explosions.
Sizing guide
EU sizes UK sizes
34,5 2
35 2.5
35,5 3
36 3.5
37 4
37,5 4.5
38 5
39 5.5
39,5 6
40 6.5
41 7
41,5 7.5
42 8
42,5 8.5
43 9
44 9.5
44,5 10
45 10.5
46 11
46,5 11.5
47 12
47,5 12.5
48 13
49 13.5
European standards for safety footwear
Safety footwear
EN ISO 20345: Highest protection. Toecaps tested to 200 joule. Classification S.
SB Basic requirements for safety footwear met - 200 joule Protection
S1 200 joule Toecap Protection. Closed seat region (fully enclosed heel). Anti-static properties. Energy absorption of seat region.
S2 200 joule Toecap Protection. Closed seat region (fully enclosed heel). Anti-static properties. Energy absorption of seat region. Water penetration and water absorption resistance.
S3 200 joule Toecap Protection. Closed seat region (fully enclosed heel). Anti-static properties. Energy absorption of seat region. Water penetration and water absorption resisitance. Plus penetration resistance. Cleated outsole.
S4 200 joule Toecap Protection. All rubber or all polymeric footwear with anti-static properties. Energy absorption of seat region.
S5 200 joule Toecap Protection. All rubber or all polymeric footwear with anti-static properties. Energy absorption of seat region. Plus penetration resistance. Cleated outsole.
Slip resistant rating - one of:
SRA Tested on ceramic tile wetted with dilute soap solution
SRB Tested on steel with glycerol
SRC SRA + SRB
Additional safety features
Should these products offer some extra protective features then the appropriate symbol will be added i.e. a product that provides 200 joule impact protection, is anti-static and has a heat resistant outsole will be marked SB A HRO.
P Penetration resistance offered by a steel midsole.
C Conductive footwear.
A Anti-static footwear.
HI Insulation against heat.
CI Insulation against cold.
E Energy absorption of seat region.
WRU Water penetration and absorption resistance.
HRO Heat resistant to 300 °C.
Protective footwear
EN ISO 20346: Lower protection. Toecaps tested to 100 joule. Classified as P.
PB Basic requirements for protective footwear met - 100 joule protection.
P1 100 joule Toecap Protection. Closed seat region (fully enclosed heel). Anti-static properties. Energy absorption of seat region.
P2 100 joule Toecap Protection. Closed seat region (fully enclosed heel). Anti-static properties. Energy absorption of seat region. Plus water penetration and water absorption resistance.
P3 100 joule Toecap Protection. Closed seat region (fully enclosed heel). Anti-static properties. Energy absorption of seat region. Water penetration and water absorption resistance. Plus penetration resistance. Cleated outsole.
P4 100 joule Toecap Protection. All rubber or all polymeric footwear with anti-static properties. Energy absorption of seat region.
P5 100 joule Toecap Protection. All rubber or all polymeric footwear with anti-static properties. Energy absorption of seat region. Plus penetration resistance. Cleated outsole.
Occupational footwear
EN ISO 20346: Lower protection. Toecaps tested to 100 joule. Classified as P.
OB Basic requirements met.
O1 Basic requirements plus: Closed seat region (fully enclosed heel). Anti-static properties. Energy absorption of seat region.
O2 Basic requirements plus: Closed seat region (fully enclosed heel). Anti-static properties. Energy absorption of seat region. Plus water penetration and absorption resistance.
O3 Basic requirements plus: Closed seat region (fully enclosed heel). Anti-static properties. Energy absorption of seat region. Water penetration and water absorption resistance. Plus penetration resistance. Cleated outsole.
O4 Basic requirements. All rubber or all polymeric footwear with anti-static properties. Energy absorption of seat region.
O5 Basic requirements. All rubber or all polymeric footwear with anti-static properties. Energy absorption of seat region. Plus penetration resistance. Cleated outsole.
ESD = Electro Static Discharge. ESD shoes protect electrostatically endangered components in sensitive workspaces and prevent sparks and explosions.
Sizing guide
EU sizes UK sizes
34,5 2
35 2.5
35,5 3
36 3.5
37 4
37,5 4.5
38 5
39 5.5
39,5 6
40 6.5
41 7
41,5 7.5
42 8
42,5 8.5
43 9
44 9.5
44,5 10
45 10.5
46 11
46,5 11.5
47 12
47,5 12.5
48 13
49 13.5