Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Volumetric flask, 10 ml, class A, conformity certified, ST10/19, ST-hollow glass stopper, Diffico blue, acc. to USP made from DURAN 1 * 10 items
Catalog # WITG3.668.012.USP
Supplier:
witeg Labortechnik
New Product
Expand image
Volumetric flask, 10 ml, class A, conformity certified, ST10/19, ST-hollow glass stopper, Diffico blue, acc. to USP made from DURAN 1 * 10 items
Catalog # WITG3.668.012.USP
Supplier:
witeg Labortechnik
New Product