MyCapLC kit containing components to assemble three complete 1/16 OD x 300 µm ID x 100 mm length UHPLC columns, with 250 mm inlet a nd 100 mm outlet tails 1/32 OD x 50 µm ID 1 * 3 items
Catalog # SGEA2251231
Supplier: Trajan Scientific and Medical