1/16 GC ferrule 0.3 mm ID (long) two-hole 85% Vespel / 15% Graphite PK10 1 * 10 items
Artikelnr. SGEA215133
Supplier: Trajan Scientific and Medical
New Product
1/16 GC ferrule 0.3 mm ID (long) two-hole 85% Vespel / 15% Graphite PK10 1 * 10 items
Artikelnr. SGEA215133
Supplier: Trajan Scientific and Medical
New Product
Frequently Bought Together
Avantor® Ultrasphere®, HPLC Columns
Supplier: Avantor
Avantor® Apollo C18, HPLC Columns
Supplier: Avantor
Avantor® Genesis, HPLC Columns, C18
Supplier: Avantor
Safety funnels with lid, 'MARCO' V2.0
Supplier: S.C.A.T.
HPLC columns, Hypercarb™
Supplier: Thermo Fisher Scientific
Guard Cartridge Holder for HPLC Columns
Supplier: Avantor
VWR® ND9, Short Thread Vials
Supplier: LA PHA PACK
Accessories for connection system GLS 80
Supplier: DWK Life Sciences
Gastight® 1000 series syringes
Supplier: HAMILTON BONADUZ
VWR® Standard Micro Syringes
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND10
Supplier: VWR Collection
dragonfly® discovery LDV reservoirs (pack of 25) 1 * 25 items
Supplier: MARKET SOURCE PART PROCESS
Grounding cable, type 23
Supplier: S.C.A.T.
Accessories for gas filters Super Clean®
Supplier: Restek
Screw caps for certified screw top vials
Supplier: Agilent
Avantor® Alltima, HPLC Columns, C18
Supplier: Avantor