P100-LMARK DLW-PTFE(B) PLUNGER PK6 (LMK.2636735-BL) 1 * 6 items
Catalog # SGEAKLMK.2636735BL
Supplier: Trajan Scientific and Medical
New Product
P100-LMARK DLW-PTFE(B) PLUNGER PK6 (LMK.2636735-BL) 1 * 6 items
Catalog # SGEAKLMK.2636735BL
Supplier: Trajan Scientific and Medical
New Product
Frequently Bought Together
ISAspher HPLC Columns, Silica
Supplier: ISERA
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
Screw caps, 9 mm, level 3 high performance applications, SureSTART™
Supplier: Thermo Fisher Scientific
ISAspher HPLC Columns, Phenyl
Supplier: ISERA
ISAspher HPLC Columns, C18
Supplier: ISERA
Capillary columns, CP-Sil 88 for FAME
Supplier: VARIAN
Crimpers and decappers, WHEATON®
Supplier: DWK Life Sciences
HPLC column tubing
Supplier: Restek
Thermoelectric flanging tools, BOLA
Supplier: Bohlender
Glass crimp top 0,3 ml microvials for <2 ml samples, level 3 high performance applications, SureSTART™
Supplier: Thermo Fisher Scientific
Starburst plugs for shell vials
Supplier: Restek
Avantor® ACE® Excel® C18-PFP, HPLC/UHPLC Columns, Analytical, 2 µm
Supplier: Avantor
SYRINGE 701 N 10µL ON-COLUMN INJECTION CEMENTED NEEDLE (32/75/3) 1 * 1 items
Supplier: HAMILTON BONADUZ
Inlet liners for Agilent GCs
Supplier: Restek
Ion pair reagents for analytical HPLC, PIC®
Supplier: WATERS
ISAspher Reversed Phase HPLC Columns, Amino
Supplier: ISERA