EMITTER TAPERED TIP, UNCOATED 360UM OD X 20UM ID X 62.5MM, 13 DEG, PK5 1 * 5 items
Catalog # SGEA063200101
Supplier: Trajan Scientific and Medical
New Product
EMITTER TAPERED TIP, UNCOATED 360UM OD X 20UM ID X 62.5MM, 13 DEG, PK5 1 * 5 items
Catalog # SGEA063200101
Supplier: Trajan Scientific and Medical
New Product
Frequently Bought Together
Air-sampler System MAS-100 NT 1 * 1 items
Supplier: Merck
LC-MS/MS system, SCIEX Triple Quad 3500
Supplier: AB SCIEX
Accessories for Aquamate Plus
Supplier: Thermo Fisher Scientific
PEEK tubing
Supplier: Trajan Scientific and Medical
HPLC tubing, stainless steel, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
UV TUBE REPLACEMENT UV-8 MODEL S 365nM 1 * 1 items
Supplier: BENDA KONRAD
Accessories for flow cups with interchangeable nozzle
Supplier: Industrial Physics
CRS SPure™ H₂O Gas Filters
Supplier: Trajan Scientific and Medical
Rotors for Cellspin III Centrifuges, Tharmac®
Supplier: THARMAC GmbH
Chromatography tubing, PFA, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
SP-5, Standard spindle 1 * 1 items
Supplier: IKA
Capillary tubing
Supplier: VICI JOUR RESEARCH
One-piece fingertight fittings
Supplier: VICI JOUR RESEARCH
Accessories for BOD measurement, mercury-free, OxiDirect
Supplier: Lovibond