BSA Conjugated Cortisone (Cor) is a purified Conjugated Small Molecules. Purity: >90% by SDS-PAGE. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Universal. Target Name: Cortisone (Cor). Target Synonyms: 17OHCS 17-OHCS 17-Hydroxy-11-D 1 * 100 µG
Catalog # MSPPRPU51259
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together