Recombinant Epstein-Barr virus Latent membrane protein 1 (LMP1), partial is a purified Recombinant Protein. Purity: >90% as determin ed by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Epstein-Barr virus (strain GD1) (HHV-4) (Human her 1 * 100 µG
Catalog # MSPPRPC20572
Supplier: MARKET SOURCE PART PROCESS