Recombinant Lymphocytic choriomeningitis virus Pre-glycoprotein polyprotein GP complex (GPC), partial is a purified Recombinant Prot ein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Lymphocytic choriom 1 * 100 µG
Catalog # MSPPRPC27107
Supplier: MARKET SOURCE PART PROCESS