ContactUsLinkComponent
Recombinant Human NACHT, LRR and PYD domains-containing protein 3 (NLRP3), partial is a purified Recombinant Protein. Purity: >90% a s determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: NLR 1 * 100 µG
  MSPPRPC25978
Recombinant Human NACHT, LRR and PYD domains-containing protein 3 (NLRP3), partial is a purified Recombinant Protein. Purity: >90% a s determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: NLR 1 * 100 µG
  MSPPRPC25978