Recombinant Epstein-Barr virus Apoptosis regulator BHRF1 (BHRF1), partial is a purified Recombinant Protein. Purity: >90% as determi ned by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Epstein-Barr virus (strain B95-8) (HHV-4) (Huma 1 * 100 µG
Catalog # MSPPRPC20441
Supplier: MARKET SOURCE PART PROCESS