Recombinant Human Tumor necrosis factor receptor superfamily member 17 protein (TNFRSF17), partial , Active Protein is a purified Ac tive Recombinant Protein. Purity: >98% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0 EU/ug as deter 1 * 100 µG
Catalog # MSPPRPC28805
Supplier: MARKET SOURCE PART PROCESS