Recombinant Rat Cystatin C (Cys-C) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0 EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Cystatin C (Cys-C). Target Synonyms: CS 1 * 100 µG
Catalog # MSPPRPU41106
Supplier: MARKET SOURCE PART PROCESS