Rat Peptidylglycine Alpha Amidating Monooxygenase (PAM), Active Protein is a purified Eukaryotic Protein. Purity: >95% by SDS-PAGE. Host: 293F Cell. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Pe 1 * 100 µG
Catalog # MSPPRPU55792
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together