Synthetic Human Mitochondrial Open Reading Frame Of The 12S rRNA-c (MOTS-c) is a purified Synthetic Peptides. Purity: >95% by SDS-PA GE. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Mitochondrial Ope 1 * 100 µG
Catalog # MSPPRPU55429
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together