Amylin Colorimetric Cell-Based ELISA Kit for detecting Amylin in researchers plate their cell line of choice. Species Reactivity: Hu man (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay 1 * 96 Tests
Catalog # MSPPEKA50030
Supplier: MARKET SOURCE PART PROCESS