MMP-14 Colorimetric Cell-Based ELISA Kit for detecting MMP-14 in researchers plate their cell line of choice. Species Reactivity: Hu man (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At leas 1 * 96 Tests
Catalog # MSPPEKA50392
Supplier: MARKET SOURCE PART PROCESS