ContactUsLinkComponent
DNA-PK Colorimetric Cell-Based ELISA Kit for detecting DNA-PK in researchers plate their cell line of choice. Species Reactivity: Hu man (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format 1 * 96 Tests
  MSPPEKA50180
DNA-PK Colorimetric Cell-Based ELISA Kit for detecting DNA-PK in researchers plate their cell line of choice. Species Reactivity: Hu man (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format 1 * 96 Tests
  MSPPEKA50180