Human OxLDL (Oxidized Low Density Lipoprotein) ELISA Kit for detecting OxLDL in serum, plasma, other biological fluids. Species Reac tivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay T 1 * 96 Tests
Catalog # MSPPEKE62539
Supplier: MARKET SOURCE PART PROCESS