Human VCAM-1/CD106 (Vascular Cell Adhesion Molecule 1) ELISA Kit for detecting VCAM-1, CD106 in serum, plasma, other biological flui ds. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.94ng/ml. Format: 96-Well 1 * 96 Tests
Catalog # MSPPEKE61454
Supplier: MARKET SOURCE PART PROCESS