Human CECR1 (Cat Eye Syndrome Chromosome Region, Candidate 1) ELISA Kit for detecting CECR1 in serum, plasma, other biological fluid s. Species Reactivity: Human (Homo sapiens). Detection Range: 0.31-20ng/ml. Sensitivity: 0.19ng/ml. Format: 96-Well P 1 * 96 Tests
Catalog # MSPPEKE61408
Supplier: MARKET SOURCE PART PROCESS