Human ALPL (Alkaline Phosphatase, Liver/Bone/Kidney) ELISA Kit for detecting ALPL in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.13-5000pg/ml. Sensitivity: 46.88pg/ml. Format: 96-Well Plate. d 1 * 96 Tests
Catalog # MSPPEKE61360
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together