Mouse OxLDL (Oxidized low-density lipoprotein) ELISA Kit for detecting OxLDL in serum, plasma, cell culture supernatant, cell or tis sue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.313-20ng/ml. Sensit 1 * 96 Tests
Catalog # MSPPEKF58134
Supplier: MARKET SOURCE PART PROCESS