Rat VLDL (Very Low Density Lipoprotein) ELISA Kit for detecting VLDL in serum, plasma, cell culture supernatant, cell or tissue lysa te, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivi 1 * 96 Tests
Catalog # MSPPEKF58614
Supplier: MARKET SOURCE PART PROCESS