Human CDK12 (Cyclin-dependent kinase 12) ELISA Kit for detecting CDK12 in cell or tissue lysate, other biological fluids. Species Re activity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. A 1 * 96 Tests
Catalog # MSPPEKF60420
Supplier: MARKET SOURCE PART PROCESS