Human E-Cad (E-Cadherin) ELISA Kit for detecting E-Cad in serum, plasma, cell culture supernatant, cell or tissue lysate, other biol ogical fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. For 1 * 96 Tests
Catalog # MSPPEKF57030
Lieferant: MARKET SOURCE PART PROCESS
Frequently Bought Together