Rat E2 (Estradiol) ELISA Kit for detecting E2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fl uids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.5-800pg/ml. Sensitivity: 7.5pg/ml. Format: 96- 1 * 96 Tests
Catalog # MSPPEKF57964
Supplier: MARKET SOURCE PART PROCESS